missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 50 Lab Coat NB212S WH
50 Lab Coat with collar, open wrists, front snap closure, 2 pockets, and serged seams for reliable protection in industrial use.
Supplier: DuPont™ ProShield™ NB212SWH4X003000
Description
- ProShield™ 50 garments protect against non-hazardous particles, light liquid splash, and aerosols in various industrial applications.
- Made with a microporous film laminated to a spunbonded polypropylene substrate for durability and comfort.
- Front snap closures allow easy donning and doffing
- Enhanced frontal protection with a laydown collar.
- Open wrists for flexible movement.
- Two front pockets for holding writing instruments, small tools, or parts.
- Manufactured without natural rubber latex.
Specifications
| 4X-Large | |
| Polypropylene, Polyethylene Laminate | |
| Snaps | |
| Single-use | |
| Non-hazardous Liquids | |
| 2 | |
| Not Autoclavable |
| Unisex | |
| White | |
| Open | |
| Non-sterile | |
| Standard Length | |
| No |