missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 50 Lab Coat NB212S WH
Description
- ProShield™ 50 garments protect against non-hazardous particles, light liquid splash, and aerosols in various industrial applications.
- Made with a microporous film laminated to a spunbonded polypropylene substrate for durability and comfort.
- Front snap closures allow easy donning and doffing
- Enhanced frontal protection with a laydown collar.
- Open wrists for flexible movement.
- Two front pockets for holding writing instruments, small tools, or parts.
- Manufactured without natural rubber latex.
Specifications
Specifications
| Size | Small |
| Gender | Unisex |
| Material | Polypropylene, Polyethylene Laminate |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Open |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | Non-hazardous Liquids |
| Style | Standard Length |
| Show More |