missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)pyridine-2-carboxaldehyde, 95%

Catalog No. p-7030955
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31858MD 250 mg
AAH3185803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31858MD Supplier Thermo Scientific Chemicals Supplier No. H31858MD

CAS: 31224-82-5 | C7H4F3NO | 175.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31224-82-5
Molecular Formula C7H4F3NO
Molecular Weight (g/mol) 175.11
MDL Number MFCD07375388
InChI Key JJDDVGAESNBKMY-UHFFFAOYSA-N
Synonym 5-trifluoromethyl picolinaldehyde, 5-trifluoromethyl-2-pyridinecarboxyaldehyde, 5-trifluoromethyl-pyridine-2-carbaldehyde, 5-trifluoromethyl pyridine-2-carboxaldehyde, 5-trifluoromethyl pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde,5-trifluoromethyl, 5-trifluoromethylpicolinealdehyde, 5-trifluoromethylpicoline aldehyde, 5-trifluoromethyl-pyridine-2-carboxaldehyde, 2-pyridinecarboxaldehyde, 5-trifluoromethyl
PubChem CID 14422792
IUPAC Name 5-(trifluoromethyl)pyridine-2-carbaldehyde
SMILES C1=CC(=NC=C1C(F)(F)F)C=O

Specifications

Melting Point 31°C to 33°C
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 175.11
Percent Purity 95%
Chemical Name or Material 5-(Trifluoromethyl)pyridine-2-carboxaldehyde
Hazard Category H302-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only