missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Trifluoromethyl-1,3-phenylenediamine 98.0+%, TCI America™
$72.70 - $359.76
Chemical Identifiers
| CAS | 368-53-6 |
|---|---|
| Molecular Formula | C7H7F3N2 |
| Molecular Weight (g/mol) | 176.142 |
| MDL Number | MFCD00014776 |
| InChI Key | KZSXRDLXTFEHJM-UHFFFAOYSA-N |
| Synonym | 3,5-diaminobenzotrifluoride, 5-trifluoromethyl benzene-1,3-diamine, 5-trifluoromethyl-1,3-phenylenediamine, 5-trifluoromethyl-1,3-benzenediamine, 3,5-benzotrifluorodiamine, unii-z89rd529kk, 5-trifluoromethyl-benzene-1,3-diamine, 1,3-benzeneamine, 5-trifluoromethyl, 1,3-benzenediamine, 5-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-3,5-diamine |
| PubChem CID | 67781 |
| IUPAC Name | 5-(trifluoromethyl)benzene-1,3-diamine |
| SMILES | C1=C(C=C(C=C1N)N)C(F)(F)F |
Chemical Identifiers
| 368-53-6 | |
| 176.142 | |
| KZSXRDLXTFEHJM-UHFFFAOYSA-N | |
| 67781 | |
| C1=C(C=C(C=C1N)N)C(F)(F)F |
| C7H7F3N2 | |
| MFCD00014776 | |
| 3,5-diaminobenzotrifluoride, 5-trifluoromethyl benzene-1,3-diamine, 5-trifluoromethyl-1,3-phenylenediamine, 5-trifluoromethyl-1,3-benzenediamine, 3,5-benzotrifluorodiamine, unii-z89rd529kk, 5-trifluoromethyl-benzene-1,3-diamine, 1,3-benzeneamine, 5-trifluoromethyl, 1,3-benzenediamine, 5-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-3,5-diamine | |
| 5-(trifluoromethyl)benzene-1,3-diamine |
Specifications
| 368-53-6 | |
| C7H7F3N2 | |
| 1 g | |
| KZSXRDLXTFEHJM-UHFFFAOYSA-N | |
| 5-(trifluoromethyl)benzene-1,3-diamine | |
| 67781 | |
| ≥98.0% (GC,T) | |
| 5-Trifluoromethyl-1,3-phenylenediamine |
| 89°C | |
| MFCD00014776 | |
| 3,5-diaminobenzotrifluoride, 5-trifluoromethyl benzene-1,3-diamine, 5-trifluoromethyl-1,3-phenylenediamine, 5-trifluoromethyl-1,3-benzenediamine, 3,5-benzotrifluorodiamine, unii-z89rd529kk, 5-trifluoromethyl-benzene-1,3-diamine, 1,3-benzeneamine, 5-trifluoromethyl, 1,3-benzenediamine, 5-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-3,5-diamine | |
| C1=C(C=C(C=C1N)N)C(F)(F)F | |
| 176.142 | |
| 176.14 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CZ1606000
TSCA : No