missing translation for 'onlineSavingsMsg'
Learn More

5-Tricosylresorcinol, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015286
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015286 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015286 Supplier Merck Emd Millipore Supplier No. 0342210MG
Only null left
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 70110-60-0
Linear Formula C29H52O2
MDL Number MFCD09743630
Synonym 1,3-Dihydroxy-5-tricosylbenzene; 5-Tricosyl-1,3-benzenediol; Tricosylresorcinol (23:0)
Type Analytical Standard
Quantity 10 mg
Formula Weight 432.72
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard