missing translation for 'onlineSavingsMsg'
Learn More

5-Thien-2-yl-1h-pyrazole-3-carboxylic acid, Thermo Scientific™

Chemical Identifiers

CAS 182415-24-3
Molecular Formula C8H6N2O2S
Molecular Weight (g/mol) 194.208
MDL Number MFCD05170066
InChI Key DUDBNCNNJMOLCC-UHFFFAOYSA-N
Synonym 5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 3-2-thienyl-1h-pyrazole-5-carboxylic acid, 5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl, 5-2-thienyl-3-pyrazolecarboxylic acid, 3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 5-2-thienyl pyrazole-3-carboxylic acid
PubChem CID 818268
IUPAC Name 5-thiophen-2-yl-1H-pyrazole-3-carboxylic acid
SMILES C1=CSC(=C1)C2=CC(=NN2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC43501DA
SDS View Documents
Thermo Scientific Chemicals
CC43501DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC43501DE
SDS View Documents
Thermo Scientific Chemicals
CC43501DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC43501CB
SDS View Documents
Thermo Scientific Chemicals
CC43501CB
250g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

182415-24-3
194.208
DUDBNCNNJMOLCC-UHFFFAOYSA-N
818268
C1=CSC(=C1)C2=CC(=NN2)C(=O)O
C8H6N2O2S
MFCD05170066
5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 3-2-thienyl-1h-pyrazole-5-carboxylic acid, 5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl, 5-2-thienyl-3-pyrazolecarboxylic acid, 3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 5-2-thienyl pyrazole-3-carboxylic acid
5-thiophen-2-yl-1H-pyrazole-3-carboxylic acid

Specifications

182415-24-3
100.0
Amber glass bottle
MFCD05170066
5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 3-2-thienyl-1h-pyrazole-5-carboxylic acid, 5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl, 5-2-thienyl-3-pyrazolecarboxylic acid, 3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 5-2-thienyl pyrazole-3-carboxylic acid
C1=CSC(=C1)C2=CC(=NN2)C(=O)O
194.208
194.21
5-Thien-2-yl-1H-pyrazole-3-carboxylic acid
96.5
97%
C8H6N2O2S
1g
DUDBNCNNJMOLCC-UHFFFAOYSA-N
5-thiophen-2-yl-1H-pyrazole-3-carboxylic acid
818268
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents