accessibility menu, dialog, popup

UserName

CAS RN 182415-24-3

OOHNNS

CAS RN 182415-24-3

IUPAC Name: 5-(thiophen-2-yl)-1H-pyrazole-3-carboxylate
Synonyms: 5-(thiophen-2-yl)-1H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 193.2
Molecular Formula: C8H5N2O2S
InChi Key: DUDBNCNNJMOLCC-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=NNC(=C1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)