missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Thiazolemethanol 98.0+%, TCI America™
Supplier: TCI America T277925G
Specifications
| 5-Thiazolemethanol | |
| Yellow | |
| C4H5NOS | |
| 25 g | |
| WKBQQWDVVHGWDB-UHFFFAOYSA-N | |
| 1,3-thiazol-5-ylmethanol | |
| 2763216 | |
| ≥98.0% (GC) |
| 38585-74-9 | |
| 106°C | |
| MFCD04115732 | |
| 5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole | |
| C1=C(SC=N1)CO | |
| 115.15 | |
| 115.15 | |
| Liquid |
Chemical Identifiers
| 38585-74-9 | |
| 115.15 | |
| WKBQQWDVVHGWDB-UHFFFAOYSA-N | |
| 2763216 | |
| C1=C(SC=N1)CO |
| C4H5NOS | |
| MFCD04115732 | |
| 5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole | |
| 1,3-thiazol-5-ylmethanol |
Safety and Handling
EINECSNumber : (5)-6483
TSCA : No
Recommended Storage : Freezer