missing translation for 'onlineSavingsMsg'
Learn More

5-Thiazolemethanol 98.0+%, TCI America™

Catalog No. p-7137412
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27795G 5 g
T277925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27795G Supplier TCI America Supplier No. T27795G
Only null left

Chemical Identifiers

CAS 38585-74-9
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
MDL Number MFCD04115732
InChI Key WKBQQWDVVHGWDB-UHFFFAOYSA-N
Synonym 5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole
PubChem CID 2763216
IUPAC Name 1,3-thiazol-5-ylmethanol
SMILES C1=C(SC=N1)CO

Specifications

Color Yellow
Boiling Point 106°C
Quantity 5 g
Formula Weight 115.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Thiazolemethanol
EINECSNumber (5)-6483
TSCA No
Recommended Storage Freezer