missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-tert-Butyl-m-xylene 98.0+%, TCI America™
$50.05 - $379.26
Chemical Identifiers
| CAS | 98-19-1 |
|---|---|
| Molecular Formula | C12H18 |
| Molecular Weight (g/mol) | 162.276 |
| MDL Number | MFCD00008832 |
| InChI Key | FZSPYHREEHYLCB-UHFFFAOYSA-N |
| Synonym | 5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl |
| PubChem CID | 7378 |
| IUPAC Name | 1-tert-butyl-3,5-dimethylbenzene |
| SMILES | CC1=CC(=CC(=C1)C(C)(C)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B096525ML
|
TCI America
B096525ML |
25 mL |
Each for $50.05
|
|
|||||
|
B0965500ML
|
TCI America
B0965500ML |
500 mL |
Each for $379.26
|
|
|||||
Chemical Identifiers
| 98-19-1 | |
| 162.276 | |
| FZSPYHREEHYLCB-UHFFFAOYSA-N | |
| 7378 | |
| CC1=CC(=CC(=C1)C(C)(C)C)C |
| C12H18 | |
| MFCD00008832 | |
| 5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl | |
| 1-tert-butyl-3,5-dimethylbenzene |
Specifications
| 98-19-1 | |
| 202°C | |
| MFCD00008832 | |
| 5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl | |
| CC1=CC(=CC(=C1)C(C)(C)C)C | |
| 162.276 | |
| 162.28 | |
| Liquid |
| Colorless | |
| C12H18 | |
| 25 mL | |
| FZSPYHREEHYLCB-UHFFFAOYSA-N | |
| 1-tert-butyl-3,5-dimethylbenzene | |
| 7378 | |
| ≥98.0% (GC) | |
| 5-tert-Butyl-m-xylene |
Safety and Handling
EINECSNumber : (3)-3427
RTECSNumber : ZE3900000
TSCA : Yes