missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl-m-xylene 98.0+%, TCI America™

Catalog No. B096525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B096525ML 25 mL
B0965500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B096525ML Supplier TCI America Supplier No. B096525ML

Chemical Identifiers

CAS 98-19-1
Molecular Formula C12H18
Molecular Weight (g/mol) 162.276
MDL Number MFCD00008832
InChI Key FZSPYHREEHYLCB-UHFFFAOYSA-N
Synonym 5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl
PubChem CID 7378
IUPAC Name 1-tert-butyl-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)C(C)(C)C)C

Specifications

Color Colorless
Boiling Point 202°C
Quantity 25 mL
Formula Weight 162.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-tert-Butyl-m-xylene
EINECSNumber (3)-3427
RTECSNumber ZE3900000
TSCA Yes