Learn More
5-tert-Butyl-m-xylene, 98%
CAS: 98-19-1 | C12H18 | 162.276 g/mol
Supplier: Thermo Scientific Chemicals A1922036
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-tert-Butyl-m-xylene | |
| -22°C | |
| 207°C to 209°C | |
| C12H18 | |
| MFCD00008832 | |
| 1853314 | |
| FZSPYHREEHYLCB-UHFFFAOYSA-N | |
| 1-tert-butyl-3,5-dimethylbenzene | |
| 7378 | |
| 98% |
| 98-19-1 | |
| 0.86 | |
| 84°C (183°F) | |
| 1.496 | |
| 500 g | |
| 5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl | |
| CC1=CC(=CC(=C1)C(C)(C)C)C | |
| 162.276 | |
| 162.27 |
Chemical Identifiers
| 98-19-1 | |
| 162.276 | |
| FZSPYHREEHYLCB-UHFFFAOYSA-N | |
| 7378 | |
| CC1=CC(=CC(=C1)C(C)(C)C)C |
| C12H18 | |
| MFCD00008832 | |
| 5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl | |
| 1-tert-butyl-3,5-dimethylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 202-647-6
RTECSNumber : ZE3900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only