missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl-m-xylene, 98%

CAS: 98-19-1 | C12H18 | 162.276 g/mol

Supplier:  Thermo Scientific Chemicals A1922022

 View more versions of this product

Catalog No. AAA1922022


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-tert-Butyl-m-xylene
-22°C
207°C to 209°C
C12H18
MFCD00008832
1853314
FZSPYHREEHYLCB-UHFFFAOYSA-N
1-tert-butyl-3,5-dimethylbenzene
7378
98%
98-19-1
0.86
84°C (183°F)
1.496
100 g
5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl
CC1=CC(=CC(=C1)C(C)(C)C)C
162.276
162.27

Chemical Identifiers

98-19-1
162.276
FZSPYHREEHYLCB-UHFFFAOYSA-N
7378
CC1=CC(=CC(=C1)C(C)(C)C)C
C12H18
MFCD00008832
5-tert-butyl-m-xylene, 1-tert-butyl-3,5-dimethylbenzene, m-xylene, 5-tert-butyl, 1,3-dimethyl-5-tert-butylbenzene, benzene, 1-1,1-dimethylethyl-3,5-dimethyl, 5-tert-butyl-1,3-dimethylbenzene, benzene, 5-tert-butyl-1,3-dimethyl, 1,3-dimethyl-5-t-butylbenzene, 5-t-butyl-1,3-dimethylbenzene, benzene, 1,3-dimethyl-5-1,1-dimethylethyl
1-tert-butyl-3,5-dimethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 202-647-6

RTECSNumber : ZE3900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only