missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl 1-Pentafluorophenyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate 98.0+%, TCI America™

Catalog No. B33185G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33185G 5 g
B33181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33185G Supplier TCI America Supplier No. B33185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86061-04-3
Molecular Formula C30H26F5NO6
Molecular Weight (g/mol) 591.531
MDL Number MFCD00065647
InChI Key AIDYQYOPUBOMTR-FQEVSTJZSA-N
Synonym fmoc-glu otbu-opfp, 5-tert-butyl 1-pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, n-fmoc-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, 5-tert-butyl1-pentafluorophenyln-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-pentafluorophenyl 5-tert-butyl ester, 1-tert-butyl 2,3,4,5,6-pentafluorophenyl 4s-4-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioate, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-tert-butyl 1-pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-t-butyl ester pentafluorphenyl ester
PubChem CID 11227121
IUPAC Name 5-O-tert-butyl 1-O-(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioate
SMILES CC(C)(C)OC(=O)CCC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 124°C
Color White
Quantity 5 g
Formula Weight 591.53
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-tert-Butyl 1-Pentafluorophenyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate
TSCA No
Recommended Storage Refrigerator