missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-SFX (6-(Fluorescein-5-Carboxamido) Hexanoic Acid, Succinimidyl Ester), single isomer

Catalog No. f6106
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F6106 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F6106 Supplier Invitrogen™ Supplier No. F6106

5-SFX (6-(Fluorescein-5-Carboxamido) Hexanoic Acid, Succinimidyl Ester), single isomer

Searching for superior alternatives to fluorescein? Our Alexa Fluor Dye Series offers everything you're looking for and more.

Specifications

Quantity 5 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.