missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-ROX, SE (5-Carboxy-X-Rhodamine, Succinimidyl Ester), single isomer

Catalog No. c6125
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C6125 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C6125 Supplier Invitrogen™ Supplier No. C6125

5-ROX, SE (5-Carboxy-X-Rhodamine, Succinimidyl Ester), single isomer

The isomeric 5-ROX, SE is an amine-reactive form of carboxy-X-rhodamine and is widely used for oligonucleotide labeling and automated DNA sequencing applications.

Specifications

Quantity 5 mg
Product Type 5-ROX SE
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye ROX
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.