missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Quinolinol 98.0+%, TCI America™
$94.33 - $296.48
Chemical Identifiers
| CAS | 578-67-6 |
|---|---|
| Molecular Formula | C9H7NO |
| Molecular Weight (g/mol) | 145.161 |
| MDL Number | MFCD00006792 |
| InChI Key | GVNQVWJYDXOLST-UHFFFAOYSA-N |
| Synonym | 5-hydroxyquinoline, quinolin-5-ol, 5-quinolinol, 5-chinolinol, ccris 4330, gyesayhwismzok-uhfffaoysa-n, 5-hydroxyquinoline 5-quinolinol, quinolin-5-one, 5-hyroxyquinoline, 5-hydroxy quinoline |
| PubChem CID | 11360 |
| IUPAC Name | 1H-quinolin-5-one |
| SMILES | C1=CC(=O)C2=CC=CNC2=C1 |
Chemical Identifiers
| 578-67-6 | |
| 145.161 | |
| GVNQVWJYDXOLST-UHFFFAOYSA-N | |
| 11360 | |
| C1=CC(=O)C2=CC=CNC2=C1 |
| C9H7NO | |
| MFCD00006792 | |
| 5-hydroxyquinoline, quinolin-5-ol, 5-quinolinol, 5-chinolinol, ccris 4330, gyesayhwismzok-uhfffaoysa-n, 5-hydroxyquinoline 5-quinolinol, quinolin-5-one, 5-hyroxyquinoline, 5-hydroxy quinoline | |
| 1H-quinolin-5-one |
Specifications
| 578-67-6 | |
| MFCD00006792 | |
| 5-hydroxyquinoline, quinolin-5-ol, 5-quinolinol, 5-chinolinol, ccris 4330, gyesayhwismzok-uhfffaoysa-n, 5-hydroxyquinoline 5-quinolinol, quinolin-5-one, 5-hyroxyquinoline, 5-hydroxy quinoline | |
| C1=CC(=O)C2=CC=CNC2=C1 | |
| 145.161 | |
| 145.16 | |
| Crystalline Powder |
| C9H7NO | |
| 1 g | |
| GVNQVWJYDXOLST-UHFFFAOYSA-N | |
| 1H-quinolin-5-one | |
| 11360 | |
| ≥98.0% (GC,T) | |
| 5-Quinolinol |
Safety and Handling
RTECSNumber : VC4100000
TSCA : No