missing translation for 'onlineSavingsMsg'
Learn More

5-Quinolinol 98.0+%, TCI America™

Catalog No. Q00591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00591G 1 g
Q00595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00591G Supplier TCI America Supplier No. Q00591G

Chemical Identifiers

CAS 578-67-6
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00006792
InChI Key GVNQVWJYDXOLST-UHFFFAOYSA-N
Synonym 5-hydroxyquinoline, quinolin-5-ol, 5-quinolinol, 5-chinolinol, ccris 4330, gyesayhwismzok-uhfffaoysa-n, 5-hydroxyquinoline 5-quinolinol, quinolin-5-one, 5-hyroxyquinoline, 5-hydroxy quinoline
PubChem CID 11360
IUPAC Name 1H-quinolin-5-one
SMILES C1=CC(=O)C2=CC=CNC2=C1

Specifications

Quantity 1 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Quinolinol
RTECSNumber VC4100000
TSCA No