missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7135240
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1759100MG 100 mg
P17591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1759100MG Supplier TCI America Supplier No. P1759100MG
Only null left

Chemical Identifiers

CAS 109299-78-7
Molecular Formula C4H5BN2O2
Molecular Weight (g/mol) 123.91
MDL Number MFCD03002366
InChI Key HZFPPBMKGYINDF-UHFFFAOYSA-N
Synonym 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid
PubChem CID 2795193
IUPAC Name (pyrimidin-5-yl)boronic acid
SMILES OB(O)C1=CN=CN=C1

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 100 mg
Formula Weight 123.91
Physical Form Crystalline Powder
Chemical Name or Material 5-Pyrimidylboronic Acid (contains varying amounts of Anhydride)
TSCA No
Recommended Storage Refrigerator