accessibility menu, dialog, popup

UserName

HOBOHNN

CAS RN 109299-78-7

IUPAC Name: (pyrimidin-5-yl)boronic acid
Synonyms: pyrimidin-5-ylboronic acid
Molecular Weight (g/mol): 123.91
Molecular Formula: C4H5BN2O2
InChi Key: HZFPPBMKGYINDF-UHFFFAOYSA-N
SMILES: OB(O)C1=CN=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)