missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidinylboronic acid, 97%, Thermo Scientific™

Catalog No. CC07412CB
Change view
Click to view available options
Quantity:
250mg
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC07412CB 250mg Amber Glass Bottle
CC07412EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC07412CB Supplier Thermo Scientific Chemicals Supplier No. CC07412CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109299-78-7
Molecular Formula C4H5BN2O2
Molecular Weight (g/mol) 123.91
MDL Number MFCD03002366,MFCD16661236
InChI Key HZFPPBMKGYINDF-UHFFFAOYSA-N
Synonym 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid
PubChem CID 2795193
IUPAC Name pyrimidin-5-ylboronic acid
SMILES OB(O)C1=CN=CN=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 123.91
Percent Purity 97%
Chemical Name or Material 5-Pyrimidinylboronic acid
Hazard Category Warning