missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Pyrimidinylboronic acid, 97%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
$638.44 - $638.44
Chemical Identifiers
| CAS | 109299-78-7 |
|---|---|
| Molecular Formula | C4H5BN2O2 |
| Molecular Weight (g/mol) | 123.91 |
| MDL Number | MFCD03002366 |
| InChI Key | HZFPPBMKGYINDF-UHFFFAOYSA-N |
| Synonym | 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid |
| PubChem CID | 2795193 |
| IUPAC Name | pyrimidin-5-ylboronic acid |
| SMILES | OB(O)C1=CN=CN=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC451700250
|
Thermo Scientific Chemicals
451700250 |
25 g | Glass bottle |
Each for $638.44
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 109299-78-7 | |
| 123.91 | |
| HZFPPBMKGYINDF-UHFFFAOYSA-N | |
| 2795193 | |
| OB(O)C1=CN=CN=C1 |
| C4H5BN2O2 | |
| MFCD03002366 | |
| 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid | |
| pyrimidin-5-ylboronic acid |
Specifications
| 109299-78-7 | |
| 96% min. (HPLC) | |
| C4H5BN2O2 | |
| 25 g | |
| HZFPPBMKGYINDF-UHFFFAOYSA-N | |
| pyrimidin-5-ylboronic acid | |
| 2795193 | |
| 97% |
| >300.0°C | |
| Glass bottle | |
| MFCD03002366 | |
| 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid | |
| OB(O)C1=CN=CN=C1 | |
| 123.91 | |
| 123.91 | |
| 5-Pyrimidinylboronic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
GHS Signal Word: Warning
RUO – Research Use Only