missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Pyridin-2-ylthiophene-2-carbaldehyde, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC02204CB
Specifications
| 5-Pyridin-2-ylthiophene-2-carbaldehyde | |
| 97% | |
| 250mg | |
| VAGVVFZWHTULBM-UHFFFAOYSA-N | |
| 5-pyridin-2-ylthiophene-2-carbaldehyde | |
| 605127 | |
| 97% |
| 132706-12-8 | |
| C10H7NOS | |
| 5-pyridin-2-yl thiophene-2-carbaldehyde, 5-2-pyridyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-carboxaldehyde, 5-2-pyridinyl-2-thiophenecarboxaldehyde, pubchem16361, acmc-1c1gm, 5-2-pyridinyl-2-thiophenecarbaldehyde, 5-2-pyridinyl-2-thiophenecarbaldehyde #, thiophene-2-carboxaldehyde, 5-2-pyridyl | |
| C1=CC=NC(=C1)C2=CC=C(S2)C=O | |
| 189.232 | |
| 189.24 |
Chemical Identifiers
| 132706-12-8 | |
| 189.232 | |
| 5-pyridin-2-yl thiophene-2-carbaldehyde, 5-2-pyridyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-carboxaldehyde, 5-2-pyridinyl-2-thiophenecarboxaldehyde, pubchem16361, acmc-1c1gm, 5-2-pyridinyl-2-thiophenecarbaldehyde, 5-2-pyridinyl-2-thiophenecarbaldehyde #, thiophene-2-carboxaldehyde, 5-2-pyridyl | |
| 5-pyridin-2-ylthiophene-2-carbaldehyde |
| C10H7NOS | |
| VAGVVFZWHTULBM-UHFFFAOYSA-N | |
| 605127 | |
| C1=CC=NC(=C1)C2=CC=C(S2)C=O |