missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Phenylvaleric Acid 99.0+%, TCI America™
Supplier: TCI America P128425G
Specifications
| 5-Phenylvaleric Acid | |
| 60°C | |
| 164°C | |
| MFCD00004416 | |
| 5-phenylvaleric acid, benzenepentanoic acid, phenylvaleric acid, phenylpentanoic acid, 5-phenylvalerate, valeric acid, 5-phenyl, 5-phenyl-pentanoic acid, 5-phenylvalericacid, unii-xyj5u8lcq8, 5-phenzylvaleric acid | |
| C1=CC=C(C=C1)CCCCC(=O)O | |
| 178.231 | |
| CHEBI:40131 | |
| ≥99.0% (T) |
| 2270-20-4 | |
| White | |
| C11H14O2 | |
| 25 g | |
| BYHDDXPKOZIZRV-UHFFFAOYSA-N | |
| 5-phenylpentanoic acid | |
| 16757 | |
| 178.23 | |
| Crystalline Powder |
Chemical Identifiers
| 2270-20-4 | |
| 178.231 | |
| BYHDDXPKOZIZRV-UHFFFAOYSA-N | |
| 16757 | |
| 5-phenylpentanoic acid |
| C11H14O2 | |
| MFCD00004416 | |
| 5-phenylvaleric acid, benzenepentanoic acid, phenylvaleric acid, phenylpentanoic acid, 5-phenylvalerate, valeric acid, 5-phenyl, 5-phenyl-pentanoic acid, 5-phenylvalericacid, unii-xyj5u8lcq8, 5-phenzylvaleric acid | |
| CHEBI:40131 | |
| C1=CC=C(C=C1)CCCCC(=O)O |
Safety and Handling
RTECSNumber : YV7816000
TSCA : No