CAS RN 2270-20-4
CAS RN 2270-20-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Phenylvaleric Acid 99.0+%, TCI America™
CAS: 2270-20-4 Molecular Formula: C11H14O2 Molecular Weight (g/mol): 178.231 MDL Number: MFCD00004416 InChI Key: BYHDDXPKOZIZRV-UHFFFAOYSA-N Synonym: 5-phenylvaleric acid,benzenepentanoic acid,phenylvaleric acid,phenylpentanoic acid,5-phenylvalerate,valeric acid, 5-phenyl,5-phenyl-pentanoic acid,5-phenylvalericacid,unii-xyj5u8lcq8,5-phenzylvaleric acid PubChem CID: 16757 ChEBI: CHEBI:40131 IUPAC Name: 5-phenylpentanoic acid SMILES: C1=CC=C(C=C1)CCCCC(=O)O