missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5-Phenylisoxazol-3-yl)methanol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 102°C |
| Color | White |
| Quantity | 1 g |
| Synonym | 5-phenylisoxazol-3-yl methanol, 5-phenyl-3-isoxazolyl methanol, 5-phenyl-1,2-oxazol-3-yl methanol, 3-isoxazolemethanol, 5-phenyl, 3-hydroxymethyl-5-phenylisoxazole, 5-phenylisoxazol-3-yl methan-1-ol, 5-phenylisoxazole-3-methanol, 3-isoxazolemethanol,5-phenyl, 5-phenyl-3-hydroxymethylisoxazole, 5-phenyl-isoxazol-3-yl methanol |
| Formula Weight | 175.19 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (5-Phenylisoxazol-3-yl)methanol |
Safety and Handling
| TSCA | No |