missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylbenzoxazole-2-thiol 98.0+%, TCI America™

Catalog No. P20111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20111G 1 g
P20115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20111G Supplier TCI America Supplier No. P20111G

Chemical Identifiers

CAS 17371-99-2
Molecular Formula C13H9NOS
Molecular Weight (g/mol) 227.28
MDL Number MFCD00022847
InChI Key QKLMWNVNJBXKRG-UHFFFAOYSA-N
Synonym 2-Mercapto-5-phenylbenzoxazole, 5-Phenylbenzoxazoline-2(3H)-thione
PubChem CID 3032663
IUPAC Name 5-phenyl-2,3-dihydro-1,3-benzoxazole-2-thione
SMILES S=C1NC2=CC(=CC=C2O1)C1=CC=CC=C1

Specifications

Melting Point 213°C
Quantity 1 g
Formula Weight 227.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenylbenzoxazole-2-thiol
RTECSNumber DM4888000
TSCA No