missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-2,4-pentadienoic acid, 98+%

Catalog No. p-7043495
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AAL0100406 5 g
AAL0100414 25 g
2 options

Catalog No. AAL0100406

Supplier: Thermo Scientific Chemicals L0100406

Only null left

CAS: 1552-94-9 | C11H10O2 | 174.199 g/mol

5-Phenyl-2,4-pentadienoic acid is used as Synthetic building block, anti-malarial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Phenyl-2,4-pentadienoic acid is used as Synthetic building block, anti-malarial agent.

Solubility
Insoluble in water.

Notes
Storage temperature: 2 -4°C and Keep container tightlyclosed in a dryand well-ventilated place.

Chemical Identifiers

CAS 1552-94-9
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD00014018
InChI Key FEIQOMCWGDNMHM-KBXRYBNXSA-N
Synonym 5-phenylpenta-2,4-dienoic acid, 2e,4e-5-phenylpenta-2,4-dienoic acid, 5-phenyl-2,4-pentadienoic acid, 2,4-pentadienoic acid, 5-phenyl, e,e-cinnamylideneacetic acid, 2e,4e-5-phenyl-2,4-pentadienoic acid, 2,4-pentadienoicacid, 5-phenyl-, 2e,4e, 4-phenyl-1,3-butadiene-1-carboxylic acid, juarezic acid, cinnamylidene acetic acid
PubChem CID 1549512
IUPAC Name (2E,4E)-5-phenylpenta-2,4-dienoic acid
SMILES C1=CC=C(C=C1)C=CC=CC(=O)O

Specifications

Melting Point 163°C to 167°C
Quantity 5 g
Beilstein 2042414
Solubility Information Insoluble in water.
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material 5-Phenyl-2,4-pentadienoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-298-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only