Learn More
5-Phenyl-2,4-pentadienoic acid, 98+%
CAS: 1552-94-9 | C11H10O2 | 174.199 g/mol
Supplier: Thermo Scientific Chemicals L0100414
Description
5-Phenyl-2,4-pentadienoic acid is used as Synthetic building block, anti-malarial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Phenyl-2,4-pentadienoic acid is used as Synthetic building block, anti-malarial agent.
Solubility
Insoluble in water.
Notes
Storage temperature: 2 -4°C and Keep container tightlyclosed in a dryand well-ventilated place.
Specifications
| 5-Phenyl-2,4-pentadienoic acid | |
| 163°C to 167°C | |
| MFCD00014018 | |
| 2042414 | |
| Insoluble in water. | |
| C1=CC=C(C=C1)C=CC=CC(=O)O | |
| 174.199 | |
| 174.2 |
| 1552-94-9 | |
| C11H10O2 | |
| 25 g | |
| 5-phenylpenta-2,4-dienoic acid, 2e,4e-5-phenylpenta-2,4-dienoic acid, 5-phenyl-2,4-pentadienoic acid, 2,4-pentadienoic acid, 5-phenyl, e,e-cinnamylideneacetic acid, 2e,4e-5-phenyl-2,4-pentadienoic acid, 2,4-pentadienoicacid, 5-phenyl-, 2e,4e, 4-phenyl-1,3-butadiene-1-carboxylic acid, juarezic acid, cinnamylidene acetic acid | |
| FEIQOMCWGDNMHM-KBXRYBNXSA-N | |
| (2E,4E)-5-phenylpenta-2,4-dienoic acid | |
| 1549512 | |
| ≥98% |
Chemical Identifiers
| 1552-94-9 | |
| 174.199 | |
| FEIQOMCWGDNMHM-KBXRYBNXSA-N | |
| 1549512 | |
| C1=CC=C(C=C1)C=CC=CC(=O)O |
| C11H10O2 | |
| MFCD00014018 | |
| 5-phenylpenta-2,4-dienoic acid, 2e,4e-5-phenylpenta-2,4-dienoic acid, 5-phenyl-2,4-pentadienoic acid, 2,4-pentadienoic acid, 5-phenyl, e,e-cinnamylideneacetic acid, 2e,4e-5-phenyl-2,4-pentadienoic acid, 2,4-pentadienoicacid, 5-phenyl-, 2e,4e, 4-phenyl-1,3-butadiene-1-carboxylic acid, juarezic acid, cinnamylidene acetic acid | |
| (2E,4E)-5-phenylpenta-2,4-dienoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-298-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only