missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1H-pyrazole-4-carboxylic acid, 97%

CAS: 5504-65-4 | C10H8N2O2 | 188.19 g/mol

Supplier:  Thermo Scientific Chemicals H3256006

 View more versions of this product

Catalog No. AAH3256006


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Phenyl-1H-pyrazole-4-carboxylic acid
280°C to 285°C
MFCD06798067,MFCD03834501
3-phenyl-1h-pyrazole-4-carboxylic acid, 3-phenylpyrazole-4-carboxylic acid, 3-phenyl-2h-pyrazole-4-carboxylic acid, 5-phenylpyrazole-4-carboxylic acid
OC(=O)C1=C(NN=C1)C1=CC=CC=C1
188.19
188.19
5504-65-4
C10H8N2O2
5 g
LGTJKUYVFSBOMC-UHFFFAOYSA-N
5-phenyl-1H-pyrazole-4-carboxylic acid
4138562
97%

Chemical Identifiers

5504-65-4
188.19
LGTJKUYVFSBOMC-UHFFFAOYSA-N
4138562
OC(=O)C1=C(NN=C1)C1=CC=CC=C1
C10H8N2O2
MFCD06798067,MFCD03834501
3-phenyl-1h-pyrazole-4-carboxylic acid, 3-phenylpyrazole-4-carboxylic acid, 3-phenyl-2h-pyrazole-4-carboxylic acid, 5-phenylpyrazole-4-carboxylic acid
5-phenyl-1H-pyrazole-4-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only