missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1H-pyrazole-4-carboxylic acid, 97%

Catalog No. p-7031472
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32560MD 250 mg
AAH3256003 1 g
AAH3256006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH32560MD Supplier Thermo Scientific Chemicals Supplier No. H32560MD
Only null left

CAS: 5504-65-4 | C10H8N2O2 | 188.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5504-65-4
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD06798067,MFCD03834501
InChI Key LGTJKUYVFSBOMC-UHFFFAOYSA-N
Synonym 3-phenyl-1h-pyrazole-4-carboxylic acid, 3-phenylpyrazole-4-carboxylic acid, 3-phenyl-2h-pyrazole-4-carboxylic acid, 5-phenylpyrazole-4-carboxylic acid
PubChem CID 4138562
IUPAC Name 5-phenyl-1H-pyrazole-4-carboxylic acid
SMILES OC(=O)C1=C(NN=C1)C1=CC=CC=C1

Specifications

Melting Point 280°C to 285°C
Quantity 250 mg
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 5-Phenyl-1H-pyrazole-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only