missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Phenyl-1-pentanol 98.0+%, TCI America™
Supplier: TCI America P132525G
Specifications
| 5-Phenyl-1-pentanol | |
| Yellow | |
| C11H16O | |
| 25 g | |
| DPZMVZIQRMVBBW-UHFFFAOYSA-N | |
| 5-phenylpentan-1-ol | |
| 61523 | |
| ≥98.0% (GC) |
| 10521-91-2 | |
| 152°C | |
| MFCD00002979 | |
| 5-phenyl-1-pentanol, benzenepentanol, 5-phenylpentanol, phenylamyl alcohol, phenylpentanol, 1-pentanol, 5-phenyl, pentanol, 5-phenyl, phenylpentanol van, unii-63ax6o7zxd, 5-phenyl-pentanol | |
| C1=CC=C(C=C1)CCCCCO | |
| 164.248 | |
| 164.25 | |
| Liquid |
Chemical Identifiers
| 10521-91-2 | |
| 164.248 | |
| DPZMVZIQRMVBBW-UHFFFAOYSA-N | |
| 61523 | |
| C1=CC=C(C=C1)CCCCCO |
| C11H16O | |
| MFCD00002979 | |
| 5-phenyl-1-pentanol, benzenepentanol, 5-phenylpentanol, phenylamyl alcohol, phenylpentanol, 1-pentanol, 5-phenyl, pentanol, 5-phenyl, phenylpentanol van, unii-63ax6o7zxd, 5-phenyl-pentanol | |
| 5-phenylpentan-1-ol |
Safety and Handling
EINECSNumber : (3)-1034
TSCA : Yes