accessibility menu, dialog, popup

UserName

CAS RN 10521-91-2

IUPAC Name: 5-phenylpentan-1-ol
Synonyms: phenylpentanol
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: DPZMVZIQRMVBBW-UHFFFAOYSA-N
SMILES: OCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)