missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1-pentanol 98.0+%, TCI America™

Catalog No. P13255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13255G 5 g
P132525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13255G Supplier TCI America Supplier No. P13255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10521-91-2
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00002979
InChI Key DPZMVZIQRMVBBW-UHFFFAOYSA-N
Synonym 5-phenyl-1-pentanol, benzenepentanol, 5-phenylpentanol, phenylamyl alcohol, phenylpentanol, 1-pentanol, 5-phenyl, pentanol, 5-phenyl, phenylpentanol van, unii-63ax6o7zxd, 5-phenyl-pentanol
PubChem CID 61523
IUPAC Name 5-phenylpentan-1-ol
SMILES C1=CC=C(C=C1)CCCCCO

Specifications

Color Yellow
Boiling Point 152°C
Quantity 5 g
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Phenyl-1-pentanol
EINECSNumber (3)-1034
TSCA Yes