missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1-pentanol, 97%

Catalog No. AAL1207003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1207003 1 g
AAL1207006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1207003 Supplier Thermo Scientific Chemicals Supplier No. L1207003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 10521-91-2 | C11H16O | 164.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 10521-91-2
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00002979
InChI Key DPZMVZIQRMVBBW-UHFFFAOYSA-N
Synonym 5-phenyl-1-pentanol, benzenepentanol, 5-phenylpentanol, phenylamyl alcohol, phenylpentanol, 1-pentanol, 5-phenyl, pentanol, 5-phenyl, phenylpentanol van, unii-63ax6o7zxd, 5-phenyl-pentanol
PubChem CID 61523
IUPAC Name 5-phenylpentan-1-ol
SMILES C1=CC=C(C=C1)CCCCCO

Specifications

Density 0.975
Boiling Point 113°C to 114°C (1.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.516
Quantity 1 g
Beilstein 1935233
Solubility Information Not miscible in water.
Formula Weight 164.25
Percent Purity 97%
Chemical Name or Material 5-Phenyl-1-pentanol
EINECSNumber 234-064-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only