missing translation for 'onlineSavingsMsg'
Learn More

5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose, Thermo Scientific Chemicals

Catalog No. AAL1598906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1598906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1598906 Supplier Thermo Scientific Chemicals Supplier No. L1598906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 141607-35-4 | Molecular Formula: C24H32O5Si | Molecular Weight: 428.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141607-35-4
Molecular Formula C24H32O5Si
Molecular Weight (g/mol) 428.6
MDL Number MFCD01075719
InChI Key OOVHEXLXHQNNLL-RQLHVLPXSA-N
PubChem CID 133612129
IUPAC Name (3aS,6S,6aS)-6-[[tert-butyl(diphenyl)silyl]oxymethyl]-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
SMILES CC1(OC2C(OC(C2O1)O)CO[Si](C3=CC=CC=C3)(C4=CC=CC=C4)C(C)(C)C)C

Specifications

Quantity 5 g
Optical Rotation −28° (c=1.5 in Chloroform)
Formula Weight 428.6
Chemical Name or Material 5-O-tert-Butyldiphenylsilyl-2,3-O-isopropylidene-alpha,beta-D-ribofuranose
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only