accessibility menu, dialog, popup

UserName

H3COOOOHOSiH3CCH3H3CH3C

CAS RN 141607-35-4

IUPAC Name: 6-{[(tert-butyldiphenylsilyl)oxy]methyl}-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-ol
Synonyms: 6-{[(tert-butyldiphenylsilyl)oxy]methyl}-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
Molecular Weight (g/mol): 428.6
Molecular Formula: C24H32O5Si
InChi Key: OOVHEXLXHQNNLL-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](OCC1OC(O)C2OC(C)(C)OC12)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results