missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-O-(tert-Butyldimethylsilyl)thymidine, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5230103
| Quantity | 1 g |
|---|
Description
5'-O-(tert-Butyldimethylsilyl)thymidine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5′-O-(tert-Butyldimethylsilyl)thymidine is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 40733-28-6 | |
| 356.49 | |
| IJWIJLIIOKZJMS-YNEHKIRRSA-N | |
| 44717046 |
| C16H28N2O5Si | |
| MFCD01631041 | |
| 1-2s,4r,5s-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione | |
| CC1=CN([C@H]2C[C@H](O)[C@@H](CO[Si](C)(C)C(C)(C)C)O2)C(=O)NC1=O |
Specifications
| 5'-O-(tert-Butyldimethylsilyl)thymidine | |
| C16H28N2O5Si | |
| MFCD01631041 | |
| IJWIJLIIOKZJMS-YNEHKIRRSA-N | |
| 1-[(2S,4R,5S)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-5-methylpyrimidine-2,4-dione | |
| 44717046 | |
| ≥97% |
| 40733-28-6 | |
| 1 g | |
| 1-2s,4r,5s-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione | |
| CC1=CN([C@H]2C[C@H](O)[C@@H](CO[Si](C)(C)C(C)(C)C)O2)C(=O)NC1=O | |
| 356.49 | |
| 356.49 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures