missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(tert-Butyldimethylsilyl)thymidine, 97+%, Thermo Scientific Chemicals

Catalog No. p-7034612
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52301MD 250 mg
AAH5230103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52301MD Supplier Thermo Scientific Chemicals Supplier No. H52301MD

5'-O-(tert-Butyldimethylsilyl)thymidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5′-O-(tert-Butyldimethylsilyl)thymidine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 40733-28-6
Molecular Formula C16H28N2O5Si
Molecular Weight (g/mol) 356.49
MDL Number MFCD01631041
InChI Key IJWIJLIIOKZJMS-YNEHKIRRSA-N
Synonym 1-2s,4r,5s-5-tert-butyldimethylsilyl oxy methyl-4-hydroxyoxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
PubChem CID 44717046
SMILES CC1=CN([C@H]2C[C@H](O)[C@@H](CO[Si](C)(C)C(C)(C)C)O2)C(=O)NC1=O

Specifications

Quantity 250 mg
IUPAC Name 1-[(2R,4S,5R)-5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Formula Weight 356.49
Percent Purity ≥97%
Chemical Name or Material 5'-O-(tert-Butyldimethylsilyl)thymidine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only