missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(4,4'-Dimethoxytrityl)thymidine 98.0+%, TCI America™

Catalog No. p-7127796
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35665G 5 g
D356625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35665G Supplier TCI America Supplier No. D35665G

Chemical Identifiers

CAS 40615-39-2
Molecular Formula C31H32N2O7
Molecular Weight (g/mol) 544.60
MDL Number MFCD00010113
InChI Key UBTJZUKVKGZHAD-QZGLRKMJNA-N
Synonym 5'-o-4,4'-dimethoxytrityl thymidine, dmt-t, thymidine, 5'-o-bis 4-methoxyphenyl phenylmethyl, 5'-o-4,4'-dimethoxytrityl thymidine dmt-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenylmethoxy methyl-4-hydroxyoxolan-2-yl-5-methylpyrimidine-2,4-dione, 5'-o-dimethoxytrityl-deoxythymidine, dmtr-dthd, dimethoxytrityl-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenyl-methoxy methyl-4-hydroxy-tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione
PubChem CID 162419
IUPAC Name 1-[(2R,4S,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C)C(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Melting Point 151°C
Color White
Quantity 5 g
Formula Weight 544.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5′-O-(4,4′-Dimethoxytrityl)thymidine
EINECSNumber (5)-6792
TSCA Yes
Recommended Storage Refrigerator