missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine, 98%

Catalog No. AAJ6558303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6558303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6558303 Supplier Thermo Scientific Chemicals Supplier No. J6558303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 114745-26-5 | C36H39N5O8 | 669.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in acetonitrile

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 114745-26-5
Molecular Formula C36H39N5O8
Molecular Weight (g/mol) 669.74
MDL Number MFCD09842119
InChI Key ISQLJOGRNUQHJX-QTXIGGTANA-N
Synonym 5'-o-dmt-n2-isobutyryl-2'-o-methyl-d-guanosine, n-9-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxy-3-methoxytetrahydrofuran-2-yl-6-oxo-6,9-dihydro-1h-purin-2-yl isobutyramide, 5'-o-dmt-n2-isobutyryl-2'-o-methyl-guanosine, n-9-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxy-3-methoxyoxolan-2-yl-6-oxo-1h-purin-2-yl-2-methylpropanamide, 5-o-dmt-n2-isobutyryl-2-o-methyl-guanosine, 5'-o-4,4'-dimethoxytrityl-n2-isobutyryl-2'-o-methylguanosine, 5'-o-bis 4-methoxyphenyl phenyl methyl-2'-o-methyl-n-2-methylpropanoyl guanosine, guanosine,5'-o-bis 4-methoxyphenyl phenylmethyl-2'-o-methyl-n-2-methyl-1-oxopropyl
PubChem CID 14991812
IUPAC Name N-[9-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxy-3-methoxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
SMILES CO[C@@H]1[C@H](O)[C@@H](COC(C2=CC=CC=C2)(C2=CC=C(OC)C=C2)C2=CC=C(OC)C=C2)O[C@H]1N1C=NC2=C1NC(NC(=O)C(C)C)=NC2=O

Specifications

Color White to Yellow
Quantity 1 g
Solubility Information Soluble in acetonitrile
Formula Weight 669.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-methylguanosine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only