missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine, 98%

Catalog No. AAJ6421706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6421706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6421706 Supplier Thermo Scientific Chemicals Supplier No. J6421706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 146954-74-7 | C30H29FN2O7 | 548.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 146954-74-7
Molecular Formula C30H29FN2O7
Molecular Weight (g/mol) 548.57
MDL Number MFCD06657648
InChI Key CSSFZSSZXOCCJB-QSZUMSSQNA-N
Synonym 1-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-3-fluoro-4-hydroxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy-2'-fluoro-uridine, 2'-deoxy-5'-o-dmt-2'-fluoro-uridine, 2'-deoxy-5'-o-dmt-2'-fluoro-d-uridine, 5'-o-bis 4-methoxyphenyl phenyl methyl-2'-deoxy-2'-fluorouridine, 5'-o-dmt-2'-fluoro-2'-deoxyuridine, 5'-o-4,4'-dimethoxytrityl-2'-fluoro-2'-deoxyuridine, uridine, 5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy-2'-fluoro, 1-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-3-fluoro-4-hydroxyoxolan-2-yl-3h-pyrimidine-2,4-dione, 2'-fluoro-5'-o-dmt-uridine
PubChem CID 7073186
IUPAC Name 1-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-3-fluoro-4-hydroxyoxolan-2-yl]pyrimidine-2,4-dione
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Color White to Yellow
Quantity 5 g
Formula Weight 548.56
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only