missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-exo,3-exo-dimethanol 97.0+%, TCI America™

Catalog No. p-7133964
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
N07691G 1 g
N07695G 5 g
2 options

Catalog No. N07691G

Supplier: TCI America N07691G

Only null left

Chemical Identifiers

CAS 699-95-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
InChI Key IGHHPVIMEQGKNE-ROVKLQMOSA-N
Synonym bicyclo 2.2.1 hept-5-ene-2-exo,3-exo-dimethanol
PubChem CID 73995587
IUPAC Name [(1R,3S,4S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol
SMILES C1C2C=CC1C(C2CO)CO

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 97°C
Quantity 1 g
Formula Weight 154.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Norbornene-2-exo,3-exo-dimethanol