CAS RN 699-95-6
IUPAC Name:
[3-(hydroxymethyl)bicyclo[2.2.1]hept-5-en-2-yl]methanol
Synonyms:
[3-(hydroxymethyl)bicyclo[2.2.1]hept-5-en-2-yl]methanol
Molecular Weight (g/mol):
154.21
Molecular Formula:
C9H14O2
InChi Key:
IGHHPVIMEQGKNE-UHFFFAOYNA-N
SMILES:
OCC1C2CC(C=C2)C1CO
1
–
2
of
2
results
1
5-Norbornene-2-exo,3-exo-dimethanol, 97%
CAS: 699-95-6 Molecular Formula: C9H14O2 Molecular Weight (g/mol): 154.209 MDL Number: MFCD00799568 InChI Key: IGHHPVIMEQGKNE-ROVKLQMOSA-N Synonym: bicyclo 2.2.1 hept-5-ene-2-exo,3-exo-dimethanol PubChem CID: 73995587 IUPAC Name: [(1R,3S,4S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol SMILES: C1C2C=CC1C(C2CO)CO
2
5-Norbornene-2-exo,3-exo-dimethanol 97.0+%, TCI America™
CAS: 699-95-6 Molecular Formula: C9H14O2 Molecular Weight (g/mol): 154.209 InChI Key: IGHHPVIMEQGKNE-ROVKLQMOSA-N Synonym: bicyclo 2.2.1 hept-5-ene-2-exo,3-exo-dimethanol PubChem CID: 73995587 IUPAC Name: [(1R,3S,4S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol SMILES: C1C2C=CC1C(C2CO)CO