missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-exo,3-exo-dimethanol 97.0+%, TCI America™

Catalog No. N07695G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07695G 5 g
N07691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07695G Supplier TCI America Supplier No. N07695G

Chemical Identifiers

CAS 699-95-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
InChI Key IGHHPVIMEQGKNE-ROVKLQMOSA-N
Synonym bicyclo 2.2.1 hept-5-ene-2-exo,3-exo-dimethanol
PubChem CID 73995587
IUPAC Name [(1R,3S,4S)-3-(hydroxymethyl)-2-bicyclo[2.2.1]hept-5-enyl]methanol
SMILES C1C2C=CC1C(C2CO)CO

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 154.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Norbornene-2-exo,3-exo-dimethanol