missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Norbornene-2-endo,3-endo-dimethanol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 86°C |
| Color | White-Yellow |
| Boiling Point | 130°C |
| Quantity | 5 g |
| Formula Weight | 154.21 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Norbornene-2-endo,3-endo-dimethanol |
Safety and Handling
| TSCA | No |