missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Norbornene-2,3-dicarboxylic Acid 98.0+%, TCI America™
$119.30 - $590.90
Chemical Identifiers
| CAS | 3813-52-3 |
|---|---|
| Molecular Formula | C9H10O4 |
| Molecular Weight (g/mol) | 182.175 |
| MDL Number | MFCD00003735 |
| InChI Key | NIDNOXCRFUCAKQ-UHFFFAOYSA-N |
| Synonym | Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Acid |
| PubChem CID | 97965 |
| IUPAC Name | bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid |
| SMILES | C1C2C=CC1C(C2C(=O)O)C(=O)O |
Chemical Identifiers
| 3813-52-3 | |
| 182.175 | |
| NIDNOXCRFUCAKQ-UHFFFAOYSA-N | |
| 97965 | |
| C1C2C=CC1C(C2C(=O)O)C(=O)O |
| C9H10O4 | |
| MFCD00003735 | |
| Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Acid | |
| bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid |
Specifications
| 3813-52-3 | |
| White | |
| MFCD00003735 | |
| Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Acid | |
| C1C2C=CC1C(C2C(=O)O)C(=O)O | |
| 182.175 | |
| 182.18 | |
| Crystalline Powder |
| 178°C | |
| C9H10O4 | |
| 5 g | |
| NIDNOXCRFUCAKQ-UHFFFAOYSA-N | |
| bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid | |
| 97965 | |
| ≥98.0% (GC,T) | |
| 5-Norbornene-2,3-dicarboxylic Acid |
Safety and Handling
TSCA : No