CAS RN 3813-52-3
CAS RN 3813-52-3
IUPAC Name:
bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
Synonyms:
bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
Molecular Weight (g/mol):
182.18
Molecular Formula:
C9H10O4
InChi Key:
NIDNOXCRFUCAKQ-UHFFFAOYNA-N
SMILES:
OC(=O)C1C2CC(C=C2)C1C(O)=O
1
–
1
of
1
results
1
5-Norbornene-2,3-dicarboxylic Acid 98.0+%, TCI America™
CAS: 3813-52-3 Molecular Formula: C9H10O4 Molecular Weight (g/mol): 182.175 MDL Number: MFCD00003735 InChI Key: NIDNOXCRFUCAKQ-UHFFFAOYSA-N Synonym: Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Acid PubChem CID: 97965 IUPAC Name: bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid SMILES: C1C2C=CC1C(C2C(=O)O)C(=O)O