missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2,2-dimethanol 98.0+%, TCI America™

Catalog No. N06361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06361G 1 g
N06365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06361G Supplier TCI America Supplier No. N06361G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6707-12-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.21
MDL Number MFCD00167595
InChI Key DSHXMENPUICESR-UHFFFAOYNA-N
Synonym Bicyclo[2.2.1]hept-5-ene-2,2-dimethanol, 2,2-Bis(hydroxymethyl)-5-norbornene
PubChem CID 110917
IUPAC Name [5-(hydroxymethyl)-5-bicyclo[2.2.1]hept-2-enyl]methanol
SMILES C1C2CC(C1C=C2)(CO)CO

Specifications

Melting Point 112°C
Quantity 1 g
Formula Weight 154.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Norbornene-2,2-dimethanol