accessibility menu, dialog, popup

UserName

CAS RN 6707-12-6

HOOH

CAS RN 6707-12-6

IUPAC Name: [2-(hydroxymethyl)bicyclo[2.2.1]hept-5-en-2-yl]methanol
Synonyms: [2-(hydroxymethyl)bicyclo[2.2.1]hept-5-en-2-yl]methanol
Molecular Weight (g/mol): 154.21
Molecular Formula: C9H14O2
InChi Key: DSHXMENPUICESR-UHFFFAOYNA-N
SMILES: OCC1(CO)CC2CC1C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results